Learn More
Research Products International Corp SPACE SAVER Polycarbonate Vacuum Desiccator with White Polypropylene Bottom, 0.31 cu. ft.
SDP
Catalog No. 50999268
Encompass
Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dry storage and protection for sensitive products.
  • Lighter and less costly than similarly sized glass models; does not chip like glass
  • Holds a full vacuum (29.9" Hg) at room temperature for 24 hours
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-999-268 Supplier Research Products International Corp Supplier No. F42025
Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dry storage and protection for sensitive products.
  • Lighter and less costly than similarly sized glass models; does not chip like glass
  • Holds a full vacuum (29.9" Hg) at room temperature for 24 hours
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.