Learn More
Braintree Scientific SpaceDrapes, 9"""" x 12"""" Rat Pouch, 5/pk, sterile
SDP
Catalog No. 501955928
Encompass
SpaceDrapes™ provide heat-retention capabilities while doubling as surgical drapes. They decrease recovery time and improve recovery prospect for animals that are under anesthesia by reducing radiant heat loss. They can be fenestrated and used as standard drapes. Available pre-packaged and pre-sterilized, or in rolls that can be sterilized by the user. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5928 Supplier Braintree Scientific Supplier No. SPDRRPS
SpaceDrapes™ provide heat-retention capabilities while doubling as surgical drapes. They decrease recovery time and improve recovery prospect for animals that are under anesthesia by reducing radiant heat loss. They can be fenestrated and used as standard drapes. Available pre-packaged and pre-sterilized, or in rolls that can be sterilized by the user. **Sold for research purposes only and not for human use**