Learn More
Starna Cells Inc SPACER
SDP
Catalog No. NC9041141
Encompass_Preferred
Cell Compartment Spacer, For 1mm path length cells, Aluminium spacers, black anodised
Catalog No. NC9041141 Supplier Starna Cells Inc Supplier No. SPA1
Cell Compartment Spacer, For 1mm path length cells, Aluminium spacers, black anodised