Learn More
Beckman Coulter SPACERS,(2) 1" BELL-TOP OPTISEAL
Catalog No. NC9745655
Encompass_Preferred
Spacer, Amber, Polyetherimide (PEI) for OptiSeal Bell-top 26x77mm Tube. Quantity: Set of 2 Spacers
Catalog No. NC9745655 Supplier Beckman Coulter Supplier No. 361669
Spacer, Amber, Polyetherimide (PEI) for OptiSeal Bell-top 26x77mm Tube. Quantity: Set of 2 Spacers