Learn More
Waters Corp Waters Spacer, 25μm
Supplier: Waters Corp 700001952
The 25μm Spacer is a small flow cell insert gasket which seals the flow cell and defines its volume.
- Functions as a small flow cell insert gasket
- Seals the flow cell
- Defines flow cell volume