Learn More
Waters Corp Waters Spacer, 50μm, Flow Cell Insert, HPLC
Catalog No. 50792653 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 50μm Spacer is a flow cell insert gasket spacer that seals the flow cell and defines flow cell volume. This size is recommended for all standard applications.
  • Seals the flow cell
  • Defines flow cell volume
  • Recommended for all standard applications
  • Associated with HPLC systems
Catalog No. 50-792-653 Supplier Waters Corp Supplier No. 700001953
The 50μm Spacer is a flow cell insert gasket spacer that seals the flow cell and defines flow cell volume. This size is recommended for all standard applications.
  • Seals the flow cell
  • Defines flow cell volume
  • Recommended for all standard applications
  • Associated with HPLC systems