Learn More
Waters Corp Waters Spacer, for μElution Plate, Positive Pressure-96 Processor
Catalog No. 50466545 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The μElution positive pressure spacer is designed for use with μElution plates. It ensures precise sample positioning by allowing μElution plates to be placed onto the spacer and then stacked onto the collection plate, maintaining the sample sequence. This spacer is compatible with μElution plates and is included with the Positive Pressure-96 Processor, also available as a spare part.
  • Used with μElution plates.
  • Preserves sample position in integrated plates.
  • Ensures consistent sample sequence.
  • Compatible with μElution plates.
  • Included with Positive Pressure-96 Processor.
  • Available as a spare part.
  • Quantity per package: 1.
Catalog No. 50-466-545 Supplier Waters Corp Supplier No. 405006528
The μElution positive pressure spacer is designed for use with μElution plates. It ensures precise sample positioning by allowing μElution plates to be placed onto the spacer and then stacked onto the collection plate, maintaining the sample sequence. This spacer is compatible with μElution plates and is included with the Positive Pressure-96 Processor, also available as a spare part.
  • Used with μElution plates.
  • Preserves sample position in integrated plates.
  • Ensures consistent sample sequence.
  • Compatible with μElution plates.
  • Included with Positive Pressure-96 Processor.
  • Available as a spare part.
  • Quantity per package: 1.