Learn More
Beckman Coulter 2 mL, Quick-Seal Bell-Top Polypropylene Tube, 11 x 32mm, 50/pk
Catalog No. NC1369260
Encompass
Quick-Seal; Polypropylene Tubes For use in Ultracentrifuge Rotors: SW 60 Ti (Part Number 335649); Required Spacer: 344674 TLA-120.2
Catalog No. NC1369260 Supplier Beckman Coulter Supplier No. 344674
Quick-Seal; Polypropylene Tubes For use in Ultracentrifuge Rotors: SW 60 Ti (Part Number 335649); Required Spacer: 344674 TLA-120.2