Learn More
Glen Research Spacer Phosphoramidite 18, 0.25 g, 18-atom hexaethylene glycol (HEG) spacer for oligonucleotide modification, Anhydrous Acetonitrile diluent
Catalog No. NC3252414
Encompass_Preferred
Spacer Phosphoramidite 18, 0.25 g, 18-atom hexaethylene glycol (HEG) spacer for oligonucleotide modification, Anhydrous Acetonitrile diluent
Catalog No. NC3252414 Supplier Glen Research Supplier No. 10191802
Spacer Phosphoramidite 18, 0.25 g, 18-atom hexaethylene glycol (HEG) spacer for oligonucleotide modification, Anhydrous Acetonitrile diluent