Learn More
Gel Company SPACER PLATE W/0.75 MM
Catalog No. NC1922813
Encompass
Spacer Plate w/0.75 mm spacer attached, for Bio-Rad Mini-PROTEAN® 3 & Tetra Cell multi-casting chamber, pkg of 5.
Catalog No. NC1922813 Supplier Gel Company Supplier No. GBU0755
Spacer Plate w/0.75 mm spacer attached, for Bio-Rad Mini-PROTEAN® 3 & Tetra Cell multi-casting chamber, pkg of 5.