Learn More
Beckman Coulter SPACERS 2/PKSPACERS 2/PK
Catalog No. NC9719284
Encompass_Preferred
Ultracentrifuge Rotor Spacers, for 8.9mL OptiSeal Tubes,.
Catalog No. NC9719284 Supplier Beckman Coulter Supplier No. 361670
Ultracentrifuge Rotor Spacers, for 8.9mL OptiSeal Tubes,.