Learn More
Hach Company SPADNS Fluoride Reagent Solution, 1 L
Supplier:  Hach Company 44453
Encompass
SPADNS Fluoride Reagent Solution. USEPA accepted for reporting for drinking and wastewater analysis (distillation required) using Hach Method 8029, Fluoride, SPADNS Method Range: 0.02 to 2.00 mg/L. 1 liter bottle
Catalog No. 50-199-4299