Learn More
Hach Company SPADNS Fluoride Reagent Solution, 3.78 L
Supplier:  Hach Company 44417
Encompass
SPADNS Fluoride Reagent Solution. USEPA accepted for reporting for drinking and wastewater analysis (distillation required) using Hach Method 8029, Fluoride, SPADNS Method Range: 0.02 to 2.00 mg/L. 3.78 liter bottle
Catalog No. 50-199-4296