Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPADNS Reference Solution, for Fluoride Determination, Ricca Chemical
SDP

Catalog No. R79900001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R79900001C 1 L
7990-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R79900001C Supplier Ricca Chemical Company Supplier No. 799032
Add to Cart
Add to Cart

APHA and EPA for Fluoride

Specifications

Chemical Name or Material SPADNS Reference Solution
Color Orange-Red
Name Note For Fluoride Determination
CAS 7732-18-5, 7647-01-0, 23647-14-5
Solubility Information Solubility in Water: Infinite
Grade APHA/EPA
Odor Slightly pungent
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
pH <1
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.