Learn More
Hach Company SPADNS 2 (Arsenic-free) Fluoride Reagent AccuVac Ampules, pk/25
Catalog No. 501996191 Shop All Hach Company Products
Encompass
For determination of Fluoride by Hach SPADNS 2 Method 10225, using AccuVac Ampules. The SPADNS 2 reagent formulation removes interference from chlorine without arsenic. EPA Compliant for use in Drinking Water and Wastewater reporting programs and equivalent to the EPA reference method SM 4500-F D. Range: 0.02 - 2.00 mg/L F- Pack of 25 ampules
Catalog No. 50-199-6191 Supplier Hach Company Supplier No. 2527025
For determination of Fluoride by Hach SPADNS 2 Method 10225, using AccuVac Ampules. The SPADNS 2 reagent formulation removes interference from chlorine without arsenic. EPA Compliant for use in Drinking Water and Wastewater reporting programs and equivalent to the EPA reference method SM 4500-F D. Range: 0.02 - 2.00 mg/L F- Pack of 25 ampules