Learn More
Hach Company SPADNS 2 (Arsenic-free) Fluoride Reagent Solution, 500 mL
Catalog No. 501996932 Shop All Hach Company Products
Encompass
For determination of Fluoride by Hach SPADNS 2 Method 10225. The SPADNS2 reagent formulation removes interference from chlorine without arsenic. Range: 0.02 - 2.00 mg/L F-. EPA compliant. 500 mL bottle
Catalog No. 50-199-6932 Supplier Hach Company Supplier No. 2947549
For determination of Fluoride by Hach SPADNS 2 Method 10225. The SPADNS2 reagent formulation removes interference from chlorine without arsenic. Range: 0.02 - 2.00 mg/L F-. EPA compliant. 500 mL bottle