Learn More
Hach Company SPADNS 2 (Arsenic-free) Fluoride Reagent Solution, 3.78 L
Catalog No. 501996931 Shop All Hach Company Products
Encompass
For determination of Fluoride by Hach SPADNS 2 Method 10225. The SPADNS2 reagent formulation removes interference from chlorine without arsenic. Range: 0.02 - 2.00 mg/L F-. EPA compliant. 3.78 liter bottle
Catalog No. 50-199-6931 Supplier Hach Company Supplier No. 2947517
For determination of Fluoride by Hach SPADNS 2 Method 10225. The SPADNS2 reagent formulation removes interference from chlorine without arsenic. Range: 0.02 - 2.00 mg/L F-. EPA compliant. 3.78 liter bottle