Learn More
Hach Company SPADNS 2 (Arsenic-free) Fluoride Reagent Solution, 1 L
Supplier:  Hach Company 2947553
Encompass
For determination of Fluoride by Hach SPADNS 2 Method 10225. The SPADNS2 reagent formulation removes interference from chlorine without arsenic. Range: 0.02 - 2.00 mg/L F-. EPA compliant. 1 liter bottle
Catalog No. 50-199-6933