Learn More
Aviva Systems Biology Corp SPAG4L Antibody - N-terminal region (ARP58530_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58530_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPAG4L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPAG4L. Alternate names: MGC33594, TSARG4, dJ726C3.1, SPAG4LApplicationWB. Species reactivity: Human
Catalog No. 50-014-62346