Learn More
Aviva Systems Biology Corp SPAG6 Antibody - middle region (ARP53681_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPAG6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPAG6. Alternate names: DKFZp434I153, MGC26276, Repro-SA-1, pf16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61087