Learn More
Medchemexpress LLC Spaglumic acid | 3106-85-2 | MFCD00133559 | 98.0% | 304.25 g·mol⁻¹ | C11H16N2O8 | 100 MG
SDP
Catalog No. 5000211509
Encompass_Preferred
Spaglumic acid (N-acetylaspartylglutamic acid, NAAG) is a naturally occurring neuropeptide found in brain tissue. Supplied as a solid analytical standard, it is intended for biochemical, neurochemical, and analytical applications that require a characterized NAAG reference material.
  • Analytical standard for neuropeptide and biochemical research.
  • High purity (98.0%) suitable for analytical use.
  • Molecular weight 304.25 g·mol⁻¹; formula C11H16N2O8.
  • Supplied as a solid, white to off-white material.
  • Available in small quantities including 100 mg for research use.
Catalog No. 50-002-11509 Supplier Medchemexpress LLC Supplier No. HY100921100MG
Spaglumic acid (N-acetylaspartylglutamic acid, NAAG) is a naturally occurring neuropeptide found in brain tissue. Supplied as a solid analytical standard, it is intended for biochemical, neurochemical, and analytical applications that require a characterized NAAG reference material.
  • Analytical standard for neuropeptide and biochemical research.
  • High purity (98.0%) suitable for analytical use.
  • Molecular weight 304.25 g·mol⁻¹; formula C11H16N2O8.
  • Supplied as a solid, white to off-white material.
  • Available in small quantities including 100 mg for research use.