Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-spaglumic acid | 4910-46-7 | MFCD00133559 | 100.0% | 304.25 g/mol | C11H16N2O8 | 5 MG
SDP

Catalog No. 5000263907
Encompass_Preferred

β-Spaglumic acid (CAS 4910-46-7) is a research chemical that functions as a NAAG peptidase inhibitor for biochemical and neuropharmacological studies. It is supplied for laboratory research use only and is not intended for human or clinical applications.

  • High purity (99.97%) suitable for analytical and pharmacological assays.
  • Soluble in water, facilitating preparation of stock solutions.
  • Molecular formula C11H16N2O8 and molecular weight 304.25 g/mol.
  • Used in studies of NAAG metabolism and neuroprotection mechanisms.
  • Available in small, precisely measured quantities for laboratory sampling.

Catalog No. 50-002-63907 Supplier Medchemexpress LLC Supplier No. HY1305535MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Spaglumic acid (CAS 4910-46-7) is a research chemical that functions as a NAAG peptidase inhibitor for biochemical and neuropharmacological studies. It is supplied for laboratory research use only and is not intended for human or clinical applications.

  • High purity (99.97%) suitable for analytical and pharmacological assays.
  • Soluble in water, facilitating preparation of stock solutions.
  • Molecular formula C11H16N2O8 and molecular weight 304.25 g/mol.
  • Used in studies of NAAG metabolism and neuroprotection mechanisms.
  • Available in small, precisely measured quantities for laboratory sampling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.