Learn More
Medchemexpress LLC Spaglumic acid (N-acetylaspartylglutamic acid) | 3106-85-2 | MFCD00076652 | 304.25 g/mol | C11H16N2O8 | 50 MG
SDP
Catalog No. 5000211588
Encompass_Preferred
Spaglumic acid (N-acetylaspartylglutamic acid) is a naturally occurring neuropeptide supplied as a solid analytical reagent for laboratory research. It is used as a reference standard in neurotransmitter studies, receptor and enzymatic assays, and analytical method development. Chemical identifiers include CAS 3106-85-2, molecular formula C11H16N2O8, and molecular weight 304.25 g/mol.
  • Provides a solid reference for biochemical and analytical assays.
  • Relevant for studies of glutamatergic neurotransmission and receptor activity.
  • Suitable for use as an analytical standard in method development.
  • Identifiable by CAS 3106-85-2 and molecular formula C11H16N2O8.
  • Solid form simplifies storage and handling in laboratory settings.
Catalog No. 50-002-11588 Supplier Medchemexpress LLC Supplier No. HY10092150MG
Add to Cart
Add to Cart
Spaglumic acid (N-acetylaspartylglutamic acid) is a naturally occurring neuropeptide supplied as a solid analytical reagent for laboratory research. It is used as a reference standard in neurotransmitter studies, receptor and enzymatic assays, and analytical method development. Chemical identifiers include CAS 3106-85-2, molecular formula C11H16N2O8, and molecular weight 304.25 g/mol.
  • Provides a solid reference for biochemical and analytical assays.
  • Relevant for studies of glutamatergic neurotransmission and receptor activity.
  • Suitable for use as an analytical standard in method development.
  • Identifiable by CAS 3106-85-2 and molecular formula C11H16N2O8.
  • Solid form simplifies storage and handling in laboratory settings.