Learn More
Novus Biologicals™ SPAM1 Synthetic Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 6677 |
| Purity | Multi-step |
| Concentration | LYOPH |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-83580. It may block above mentioned antibody from binding to its target protein in western blot and/or immunohistochemistry under proper experimental settings. |
| Gene Alias | EC 3.2.1.35, HYA1, HYAL1, HYAL3, HYAL5, Hyal-PH20, hyaluronidase PH-20, Hyaluronoglucosaminidase PH-20, MGC26532, PH20, PH-20, SPAG15, Sperm adhesion molecule 1, sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding), Sperm surface protein PH-20 |
| Gene Symbol | SPAM1 |
| Molecular Weight (g/mol) | M.W. Theoretical: 55 kDa |
| Quantity | 100 μg |
| Research Category | Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.