Learn More
Beckman Coulter Biomek Span-8 P250 Conductive Tips
Catalog No. NC0268300 Shop All Beckman Coulter Products
Encompass
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing
Catalog No. NC0268300 Supplier Beckman Coulter Supplier No. 719011
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing