Learn More
Sigma Aldrich Fine Chemicals Biosciences Span 80, 1338-43-8, MFCD00080948, 250mL
Catalog No. NC1615941
Encompass
Empirical Formula (Hill Notation) C24H44O6, Molecular Weight 428.60, viscosity 1000-2000 mPa.s (20 °C), Synonym: Sorbitan monooleate, Span 80; pan 80 is a sorbitan ester series of surfactant. It is produced by esterification of one or more sorbitan hydroxyl group with fatty acid. It is hydrophobic in nature.
Catalog No. NC1615941 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 85548250ML
Empirical Formula (Hill Notation) C24H44O6, Molecular Weight 428.60, viscosity 1000-2000 mPa.s (20 °C), Synonym: Sorbitan monooleate, Span 80; pan 80 is a sorbitan ester series of surfactant. It is produced by esterification of one or more sorbitan hydroxyl group with fatty acid. It is hydrophobic in nature.