Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Span™ 80 for GC, MilliporeSigma™ Supelco™

Catalog No. 111012965 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2965 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2965 Supplier MilliporeSigma Supelco Supplier No. 0956950ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00080948
Synonym Sorbitan monooleate; Span(R) 80

Specifications

Density 0.99 g/mL at 20°C; 0.986 g/mL (at 25°C)
CAS 1338-43-8
Quantity 50 mL
Flash Point >113°C
Linear Formula C24H44O6
Formula Weight 428.60
Grade GC
RTECSNumber WG2932400

Span is a registered trademark of Croda International PLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.