Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Spanish Double RTK Compliance Center with MSDS Binders

Catalog No. 19103281 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Information Center
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-103-281 Information Center
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-281 Supplier Brady™ Supplier No. 2010DBSP

missing translation for 'validMainframeMsg'

Safety information centers provide a centralized location for important safety information and makes the information accessible to employees.

  • This Right-to-Know Compliance Center helps you comply with OSHA requirements by keeping important Material Safety Data Sheets (MSDS) easily accessible
  • Bright red and yellow board makes the information center highly visible to employees
  • Helps prevent costly mistakes by providing potentially life-saving information to employees
  • Ideal for improving awareness of your company's safety information

Specifications

Format Wall Mounted
Product Type Information Center
Includes (2) Binder Racks; (2) MSDS Binders; Backboard
Quantity Each
For Use With (Application) Hazard Communication
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.