Learn More
Enzo Life Sciences Spantide 3 (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP5040001
Encompass_Preferred
A superior substance P antagonist with high potency and negligible neurotoxicity. More effective than spantide II. Purity: ≥98% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1355