Learn More
Medchemexpress LLC Spantide I (TFA) | 99.32% | 5 MG
SDP
Catalog No. 5000424564
Encompass_Preferred
Spantide I TFA is a substance P analog and a selective NK1 receptor antagonist. It has Ki values of 230 nM for NK1 and 8150 nM for NK2 receptors. This compound can reduce type 1 cytokines and enhance type 2 cytokine IL-10 in infected corneas, which leads to a significant reduction in corneal perforation. It is for research use only, not for sale to patients.
  • Substance P analog
  • Selective NK1 receptor antagonist
  • Reduces type 1 cytokines
  • Enhances type 2 cytokine IL-10
  • Reduces corneal perforation
Catalog No. 50-004-24564 Supplier Medchemexpress LLC Supplier No. HYP1194A5MG
Spantide I TFA is a substance P analog and a selective NK1 receptor antagonist. It has Ki values of 230 nM for NK1 and 8150 nM for NK2 receptors. This compound can reduce type 1 cytokines and enhance type 2 cytokine IL-10 in infected corneas, which leads to a significant reduction in corneal perforation. It is for research use only, not for sale to patients.
  • Substance P analog
  • Selective NK1 receptor antagonist
  • Reduces type 1 cytokines
  • Enhances type 2 cytokine IL-10
  • Reduces corneal perforation