Learn More
Aviva Systems Biology Corp SPANXC Antibody - N-terminal region (ARP59900_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59900_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPANXC. It was validated on Western Blot. . Alternate names: C, CT11.3, CTp11, SPANX-CApplicationWB. Species reactivity: Human
Catalog No. 50-286-871