Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SPARC Human HEK293 100ug
SDP

Catalog No. 5000196659
Encompass_Preferred

Recombinant human SPARC protein expressed in HEK293 cells with a C-terminal 6xHis tag. Lyophilized protein supplied at 100 μg for research use; characterized by greater than 95% purity and a glycosylation-dependent molecular weight (approximately 37-46 kDa). Store at -20°C and aliquot for long-term storage.

  • Expressed in HEK293 cells with a C-terminal His tag.
  • Full-length mature protein, accession P09486 (A18-I303).
  • Molecular weight approximately 37-46 kDa due to glycosylation.
  • Formulated lyophilized with trehalose for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Recommended storage at -20°C; freeze aliquots for long-term storage.

Catalog No. 50-001-96659 Supplier Medchemexpress LLC Supplier No. HYP71094100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SPARC protein expressed in HEK293 cells with a C-terminal 6xHis tag. Lyophilized protein supplied at 100 μg for research use; characterized by greater than 95% purity and a glycosylation-dependent molecular weight (approximately 37-46 kDa). Store at -20°C and aliquot for long-term storage.

  • Expressed in HEK293 cells with a C-terminal His tag.
  • Full-length mature protein, accession P09486 (A18-I303).
  • Molecular weight approximately 37-46 kDa due to glycosylation.
  • Formulated lyophilized with trehalose for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Recommended storage at -20°C; freeze aliquots for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.