Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SPARC Human HEK293 5ug
SDP

Catalog No. 5000196735
Encompass_Preferred

Recombinant human SPARC (secreted protein acidic and rich in cysteine) produced in HEK293 mammalian cells and supplied as a lyophilized, C-terminal His-tagged protein for biochemical and cell-based studies involving extracellular matrix interactions and lipoprotein binding. Suitable for binding assays, structural studies, and functional assays.

  • Expressed in HEK293 mammalian cells for native glycosylation.
  • C-terminal 6xHis tag enabling affinity purification and detection.
  • Available in multiple sizes down to 5 μg for small-scale experiments.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for stability and long-term storage.
  • Includes full sequence corresponding to UniProt residues A18-I303 and glycosylated molecular weight ~37-46 kDa.

Catalog No. 50-001-96735 Supplier Medchemexpress LLC Supplier No. HYP710945UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SPARC (secreted protein acidic and rich in cysteine) produced in HEK293 mammalian cells and supplied as a lyophilized, C-terminal His-tagged protein for biochemical and cell-based studies involving extracellular matrix interactions and lipoprotein binding. Suitable for binding assays, structural studies, and functional assays.

  • Expressed in HEK293 mammalian cells for native glycosylation.
  • C-terminal 6xHis tag enabling affinity purification and detection.
  • Available in multiple sizes down to 5 μg for small-scale experiments.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for stability and long-term storage.
  • Includes full sequence corresponding to UniProt residues A18-I303 and glycosylated molecular weight ~37-46 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.