Learn More
Medchemexpress LLC SPARC Mouse HEK293 100ug
SDP
Catalog No. 5000196500
Encompass_Preferred
Recombinant full-length mouse SPARC protein produced in HEK293 cells with a C-terminal 6xHis tag, provided for use in biochemical and cell-based assays probing extracellular matrix interactions and SPARC-related signaling. Documentation includes sequence, purity, and storage recommendations to support reproducible experiments.
  • Expressed in HEK293 cells for appropriate post-translational modifications.
  • C-terminal 6xHis tag enables efficient affinity purification.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Includes sequence and accession information for reproducibility.
  • Stable for long-term frozen storage; use carrier protein recommendations for extended stability.
Catalog No. 50-001-96500 Supplier Medchemexpress LLC Supplier No. HYP71086100UG
Recombinant full-length mouse SPARC protein produced in HEK293 cells with a C-terminal 6xHis tag, provided for use in biochemical and cell-based assays probing extracellular matrix interactions and SPARC-related signaling. Documentation includes sequence, purity, and storage recommendations to support reproducible experiments.
  • Expressed in HEK293 cells for appropriate post-translational modifications.
  • C-terminal 6xHis tag enables efficient affinity purification.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Includes sequence and accession information for reproducibility.
  • Stable for long-term frozen storage; use carrier protein recommendations for extended stability.