Learn More
Medchemexpress LLC SPARC Mouse HEK293 10ug
SDP
Catalog No. 5000196769
Encompass_Preferred
Recombinant mouse SPARC protein (residues A18-I302) produced in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. Provided at high purity for biochemical and cell biology applications, the product includes datasheet and COA information with recommended reconstitution and storage conditions.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag for affinity purification and detection.
  • High purity (≥98%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 37-43 kDa due to glycosylation.
  • Lyophilized formulation from PBS, pH 7.4, with optional trehalose for stability.
  • Available in multiple sizes from 5 μg to 1 mg to suit assay needs.
  • Reconstitute at ≥100 μg/mL and store aliquots at -20°C or -80°C for long-term storage.
Catalog No. 50-001-96769 Supplier Medchemexpress LLC Supplier No. HYP7108610UG
Recombinant mouse SPARC protein (residues A18-I302) produced in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. Provided at high purity for biochemical and cell biology applications, the product includes datasheet and COA information with recommended reconstitution and storage conditions.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag for affinity purification and detection.
  • High purity (≥98%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 37-43 kDa due to glycosylation.
  • Lyophilized formulation from PBS, pH 7.4, with optional trehalose for stability.
  • Available in multiple sizes from 5 μg to 1 mg to suit assay needs.
  • Reconstitute at ≥100 μg/mL and store aliquots at -20°C or -80°C for long-term storage.