Learn More
Medchemexpress LLC SPARC Mouse HEK293 5ug
SDP
Catalog No. 5000196109
Encompass_Preferred
Recombinant mouse SPARC protein expressed in HEK293 cells with a His tag, provided as purified protein for research use in studies of extracellular matrix interactions and regulation of cell growth.
  • HEK293 expression ensures mammalian post-translational modifications.
  • His tag facilitates purification and detection.
  • Supplied in multiple sizes for experimental flexibility.
  • Approximate molecular weight 37-43 kDa due to glycosylation.
  • High purity suitable for biochemical and cell-based assays.
Catalog No. 50-001-96109 Supplier Medchemexpress LLC Supplier No. HYP710865UG
Recombinant mouse SPARC protein expressed in HEK293 cells with a His tag, provided as purified protein for research use in studies of extracellular matrix interactions and regulation of cell growth.
  • HEK293 expression ensures mammalian post-translational modifications.
  • His tag facilitates purification and detection.
  • Supplied in multiple sizes for experimental flexibility.
  • Approximate molecular weight 37-43 kDa due to glycosylation.
  • High purity suitable for biochemical and cell-based assays.