Learn More
ABclonal Technology SPARC Rabbit pAb
SDP
Supplier:  ABclonal Technology A144941000UL
Encompass_Preferred
This gene encodes a cysteine-rich acidic matrix-associated protein. The encoded protein is required for the collagen in bone to become calcified but is also involved in extracellular matrix synthesis and promotion of changes to cell shape. The gene product has been associated with tumor suppression but has also been correlated with metastasis based on changes to cell shape which can promote tumor cell invasion. Three transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-07616