Learn More
Medchemexpress LLC SPARC Rat HEK293 10ug
SDP
Catalog No. 5000180559
Encompass_Preferred
SPARC protein is involved in the regulation of cell growth and may play a regulatory role by affecting the interaction of extracellular matrix and cytokines SPRC protein can bind calcium and copper several types of collagen albumin thrombospondin PDGF and cell membranes It has two calcium binding sites one is an acidic domain that can bind 5-8 Ca2 with low affinity and the other is an EF-hand loop that can bind Ca2 ions with high affinity SPARC Protein Rat (HEK293 His) is the recombinant rat-derived SPARC protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80559 Supplier Medchemexpress LLC Supplier No. HYP7342010UG
SPARC protein is involved in the regulation of cell growth and may play a regulatory role by affecting the interaction of extracellular matrix and cytokines SPRC protein can bind calcium and copper several types of collagen albumin thrombospondin PDGF and cell membranes It has two calcium binding sites one is an acidic domain that can bind 5-8 Ca2 with low affinity and the other is an EF-hand loop that can bind Ca2 ions with high affinity SPARC Protein Rat (HEK293 His) is the recombinant rat-derived SPARC protein expressed by HEK293 with C-His labeled tag