Learn More
Enterprise Technology Solutions Sparco Gaffer Tape, Matte Finish, 2" x 60 Yards, 1/RL, Black
SDP
Catalog No. 5000826847
Encompass_Preferred
Gaffer tape features a low gloss, matte finish to minimize bright lights from reflecting off the surface. Strong adhesive bonds securely to surfaces to prevent peeling at the corners. Gaffer tape is perfect for bundling and wrapping wires, holding wires/cords to prevent trip hazards, masking stage floors, masking onscenery panels or backstage areas, and more.
Catalog No. 50-008-26847 Supplier Enterprise Technology Solutions Supplier No. SPR64019
Gaffer tape features a low gloss, matte finish to minimize bright lights from reflecting off the surface. Strong adhesive bonds securely to surfaces to prevent peeling at the corners. Gaffer tape is perfect for bundling and wrapping wires, holding wires/cords to prevent trip hazards, masking stage floors, masking onscenery panels or backstage areas, and more.