Learn More
Enterprise Technology Solutions Sparco Packaging Tape, 3" Core, 3.0 mil, 1.88"x55 Yds, 1 Roll, CL
SDP
Catalog No. 5000827303
Encompass_Preferred
Heavy-duty packaging tape with acrylic adhesive provides a firm, strong bond. Acrylic-based adhesive bonds well to polar surfaces like metal, glass, polyesters and polycarbonates. It also offers resistance to excellent temperature, UV and oxidation. Tape resists tearing, splitting and breakage. Design includes a 3" core and 3.0 mil thickness.
Catalog No. 50-008-27303 Supplier Enterprise Technology Solutions Supplier No. SPR64030
Heavy-duty packaging tape with acrylic adhesive provides a firm, strong bond. Acrylic-based adhesive bonds well to polar surfaces like metal, glass, polyesters and polycarbonates. It also offers resistance to excellent temperature, UV and oxidation. Tape resists tearing, splitting and breakage. Design includes a 3" core and 3.0 mil thickness.