Learn More
Enterprise Technology Solutions Sparco School Glue, 1.25oz., 12/BX, White
SDP
Catalog No. 5000826738
Encompass_Preferred
All-purpose, premium-quality glue features a no-run formula that is great for use on vertical surfaces. The glue stays put instead of dripping down and ruining your project. It dries clear and strongly bonds on most porous surfaces such as paper, cloth and semi-porous materials such as wood and pottery. It is adjustable before setting. Nontoxic glue is also washable for easier cleanup. The 1.25 oz bottle with a twist-off cap is perfect for student school boxes. Glue is not recommended for photos, bare metal and submerged or heated surfaces.
Catalog No. 50-008-26738 Supplier Enterprise Technology Solutions Supplier No. SPR15158
All-purpose, premium-quality glue features a no-run formula that is great for use on vertical surfaces. The glue stays put instead of dripping down and ruining your project. It dries clear and strongly bonds on most porous surfaces such as paper, cloth and semi-porous materials such as wood and pottery. It is adjustable before setting. Nontoxic glue is also washable for easier cleanup. The 1.25 oz bottle with a twist-off cap is perfect for student school boxes. Glue is not recommended for photos, bare metal and submerged or heated surfaces.