Learn More
Agilent Technologies Spare AMX Aspirate Manifold
GREENER_CHOICE
Catalog No. 1260552S Shop All Agilent Technologies Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
12-605-52S Autoclavable
1 options
Catalog No. 12-605-52S Supplier Agilent Technologies Supplier No. 1260552S
Add to Cart
Add to Cart
Spare Automated Media Exchange (AMX) Module Aspirate Manifold for Agilent BioTek MultiFlo FX Multimode Dispenser.
  • Spare dispense manifold (also shown in photos) purchased separately
TRUSTED_SUSTAINABILITY
Specifications
Product Type Aspirate Manifold
Autoclavable Autoclavable