Learn More
World Precision Instrument Spare Parts Kit for Nanoliter 2010
SDP
Supplier:  World Precision Instrument 300521
Encompass
  • Includes: MicroFil 34g, displacement piston, and five O-ring sets
  • For use with Nanoliter2010
  • Not compatible with Nanoliter2000
Catalog No. 50-252-1484