Learn More
World Precision Instrument Spare Parts Kit For Nanoliter2020
SDP
Catalog No. 502521474
Encompass
The Spare Parts Kits for NANOLITER2020 includes the following: 
Allen Wrench 0.035 Hex Tool
MicroFil 34 ga
O-Ring Kit for NANOLITER2020
1 cc Syringe
Replacement Piston for NANOLITER2020
Catalog No. 50-252-1474 Supplier World Precision Instrument Supplier No. 300746
The Spare Parts Kits for NANOLITER2020 includes the following: 
Allen Wrench 0.035 Hex Tool
MicroFil 34 ga
O-Ring Kit for NANOLITER2020
1 cc Syringe
Replacement Piston for NANOLITER2020