Learn More
Electron Microscopy Sciences BioCulture™ Microbial Air Sampler Spare Sampling Head for BioCulture Sampler
SDP
Catalog No. 5033877
Encompass
Sampling head design of 380 1mm diameter holes. Sampling head may be sterilized by autoclave or gas torch.
Catalog No. 50-338-77 Supplier Electron Microscopy Sciences Supplier No. 6522455
Sampling head design of 380 1mm diameter holes. Sampling head may be sterilized by autoclave or gas torch.