Learn More
Perkin Elmer US LLC Spare seaFAST Trace Metal Cleanup Column
Catalog No. 502105193
Encompass_Preferred
This Brine Concentration Column with connection lines has a 0.5mm I.D. and connects to V1 ports #1 and #4. It connects between the buffer mixing tee and buffer probe.
Catalog No. 50-210-5193 Supplier Perkin Elmer US LLC Supplier No. N0777470
This Brine Concentration Column with connection lines has a 0.5mm I.D. and connects to V1 ports #1 and #4. It connects between the buffer mixing tee and buffer probe.