Learn More
Eppendorf 3P SPARE SEALING, 5/PK
Catalog No. NC1461578
Encompass_Preferred
Spare sealing, for 750 mL round buckets, for Rotor S-4-104, 5 pcs.
Catalog No. NC1461578 Supplier Eppendorf 3P Supplier No. 5820749008
Spare sealing, for 750 mL round buckets, for Rotor S-4-104, 5 pcs.