Learn More
World Precision Instrument Spare Silicone Gasket for NanoFil & Holder
SDP
Catalog No. NC1287430
Encompass
This special silicone gasket allows metal tips, glass, and quartz tubing to be held securely in the syringe and the injection holder
Catalog No. NC1287430 Supplier World Precision Instrument Supplier No. NFGSK5
This special silicone gasket allows metal tips, glass, and quartz tubing to be held securely in the syringe and the injection holder