Learn More
Waters Corp Spares Kit, for ACQUITY RDa System, MS Type
Catalog No. 502390632 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Spares Kit is designed for an ACQUITY RDa system. It contains various replacement components for maintenance.
  • Aperture disks
  • O-ring removal tool
  • Flangeless nut tool
  • O-rings
  • Fittings
  • PEEK tubing kit
Catalog No. 50-239-0632 Supplier Waters Corp Supplier No. 700012101
This Spares Kit is designed for an ACQUITY RDa system. It contains various replacement components for maintenance.
  • Aperture disks
  • O-ring removal tool
  • Flangeless nut tool
  • O-rings
  • Fittings
  • PEEK tubing kit