Learn More
Waters Corp Waters Spares Kit, Reference Probe Assembly, MS System Type, 1 Unit per Package
Catalog No. 50466610 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Probe Assembly Spares Kit contains all the consumable parts required to maintain the reference probe on a regular basis.
  • Spare parts kit
  • For MS system types
  • Replaced as required
  • One unit per package
Catalog No. 50-466-610 Supplier Waters Corp Supplier No. 700004596
The Probe Assembly Spares Kit contains all the consumable parts required to maintain the reference probe on a regular basis.
  • Spare parts kit
  • For MS system types
  • Replaced as required
  • One unit per package